Product Details

SNP ID
rs555432
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:91262645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACGGCAAACAAGAAAAACCATT[C/T]TAATTTGGATCATAATGATTTTTGG
Phenotype
MIM: 615684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HFM1 PubMed Links

Gene Details

Gene
HFM1
Gene Name
HFM1, ATP dependent DNA helicase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017975.4 Intron NP_001017975.4
XM_011540849.1 Intron XP_011539151.1
XM_011540850.2 Intron XP_011539152.1
XM_011540851.1 Intron XP_011539153.1
XM_011540852.2 Intron XP_011539154.1
XM_011540855.1 Intron XP_011539157.1
XM_011540857.1 Intron XP_011539159.1
XM_011540859.2 Intron XP_011539161.1
XM_017000490.1 Intron XP_016855979.1
XM_017000491.1 Intron XP_016855980.1
XM_017000492.1 Intron XP_016855981.1
XM_017000493.1 Intron XP_016855982.1
XM_017000494.1 Intron XP_016855983.1
XM_017000495.1 Intron XP_016855984.1
XM_017000496.1 Intron XP_016855985.1

View Full Product Details