Product Details

SNP ID
rs344357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1786254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCACGAAGGAACGCATGCACCCTT[C/G]CACACAGCGAGAGGAGTTGGGCAGT
Phenotype
MIM: 610886 MIM: 601489 MIM: 610779 MIM: 611659
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EME2 PubMed Links

Gene Details

Gene
EME2
Gene Name
essential meiotic structure-specific endonuclease subunit 2
There are no transcripts associated with this gene.

Gene
IGFALS
Gene Name
insulin like growth factor binding protein acid labile subunit
There are no transcripts associated with this gene.

Gene
NUBP2
Gene Name
nucleotide binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284501.1 Intron NP_001271430.1
NM_001284502.1 Intron NP_001271431.1
NM_012225.3 Intron NP_036357.1
XM_005255027.2 Intron XP_005255084.1
XM_005255030.1 Intron XP_005255087.1
XM_011522338.2 Intron XP_011520640.1
XM_017022831.1 Intron XP_016878320.1
XM_017022832.1 Intron XP_016878321.1
Gene
SPSB3
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
There are no transcripts associated with this gene.

View Full Product Details