Product Details

SNP ID
rs129336
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:36759413 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGAATTTTTGTTACCTGAGAGAGA[A/C]AAGAGAAGTTCAGGAACTTGCCCTA
Phenotype
MIM: 615870
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFT27 PubMed Links

Gene Details

Gene
IFT27
Gene Name
intraflagellar transport 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177701.2 Intron NP_001171172.1
NM_006860.4 Intron NP_006851.1
XM_006724106.2 Intron XP_006724169.1
XM_017028540.1 Intron XP_016884029.1
Gene
LOC105373021
Gene Name
uncharacterized LOC105373021
There are no transcripts associated with this gene.

View Full Product Details