Product Details

SNP ID
rs591728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100714748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTTAAACAAAGACAAAAGTGTT[C/T]CTTTATTGTAGGAGTTTTCAGATTA
Phenotype
MIM: 602498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFG PubMed Links

Gene Details

Gene
TFG
Gene Name
TRK-fused gene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007565.2 Intron NP_001007566.1
NM_001195478.1 Intron NP_001182407.1
NM_001195479.1 Intron NP_001182408.1
NM_006070.5 Intron NP_006061.2
XM_005247066.1 Intron XP_005247123.1
XM_006713472.1 Intron XP_006713535.1
XM_006713473.1 Intron XP_006713536.1
XM_011512334.1 Intron XP_011510636.1
XM_017005527.1 Intron XP_016861016.1
XM_017005528.1 Intron XP_016861017.1
XM_017005529.1 Intron XP_016861018.1
XM_017005530.1 Intron XP_016861019.1

View Full Product Details