Product Details

SNP ID
rs635652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197795268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATTATTCAGTCCCTTCTGGTAA[C/T]AGTACTCCAATTTTCCTAGGGAACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRCH3 PubMed Links

Gene Details

Gene
LRCH3
Gene Name
leucine rich repeats and calponin homology domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032773.3 Intron NP_116162.1
XM_005269362.2 Intron XP_005269419.1
XM_005269363.2 Intron XP_005269420.1
XM_005269365.2 Intron XP_005269422.1
XM_005269367.2 Intron XP_005269424.1
XM_006713791.3 Intron XP_006713854.1
XM_006713792.3 Intron XP_006713855.1
XM_006713793.2 Intron XP_006713856.1
XM_011513242.2 Intron XP_011511544.1
XM_011513243.2 Intron XP_011511545.1
XM_017007350.1 Intron XP_016862839.1
XM_017007351.1 Intron XP_016862840.1
XM_017007352.1 Intron XP_016862841.1

View Full Product Details