Product Details

SNP ID
rs683227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42213328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCAAAGTGGAAGTTTGGACTAG[C/T]AAGCATGAGCTCACTACAGCCGCTC
Phenotype
MIM: 612188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM87A PubMed Links

Gene Details

Gene
TMEM87A
Gene Name
transmembrane protein 87A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110503.2 Intron NP_001103973.1
NM_001286487.1 Intron NP_001273416.1
NM_015497.4 Intron NP_056312.2
XM_005254287.1 Intron XP_005254344.1
XM_006720468.2 Intron XP_006720531.1
XM_011521443.2 Intron XP_011519745.1
XM_011521444.2 Intron XP_011519746.1
XM_017022067.1 Intron XP_016877556.1
Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
There are no transcripts associated with this gene.

View Full Product Details