Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001172698.1 | Intron | NP_001166169.1 | |||
NM_001172699.1 | Intron | NP_001166170.1 | |||
NM_133263.3 | Intron | NP_573570.3 | |||
XM_005268372.4 | Intron | XP_005268429.1 | |||
XM_011537553.2 | Intron | XP_011535855.1 | |||
XM_011537554.2 | Intron | XP_011535856.1 | |||
XM_011537555.2 | Intron | XP_011535857.1 | |||
XM_011537556.2 | Intron | XP_011535858.1 | |||
XM_011537557.1 | Intron | XP_011535859.1 |