Product Details

SNP ID
rs104893786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129528777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAACTTCTACGTGCCCTTCTCCA[A/G]TGCGACGGGTGTGGTACGCAGCCCC
Phenotype
MIM: 606045 MIM: 180380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT122 PubMed Links

Gene Details

Gene
IFT122
Gene Name
intraflagellar transport 122
There are no transcripts associated with this gene.

Gene
RHO
Gene Name
rhodopsin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000539.3 139 Missense Mutation AAT,AGT N15S NP_000530.1

View Full Product Details