Product Details

SNP ID
rs291100
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:206931468 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCCCTCATGCTGCATTTCTTTGTCA[C/T]GTAGTGGGGCTTCCTTCTTCCTCCT
Phenotype
MIM: 606015 MIM: 173880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
PIGR
Gene Name
polymeric immunoglobulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002644.3 Intron NP_002635.2
XM_011509629.1 Intron XP_011507931.1

View Full Product Details