Product Details

SNP ID
rs333237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:107987503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTAGTTGCTATAGTAACTTGAA[C/T]TACAGAAGCTTGTTGAAAATGTGGA
Phenotype
MIM: 608761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC5A7 PubMed Links

Gene Details

Gene
SLC5A7
Gene Name
solute carrier family 5 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305005.2 534 Intron NP_001291934.1
NM_001305006.2 534 Intron NP_001291935.1
NM_001305007.2 534 Intron NP_001291936.1
NM_021815.4 534 Intron NP_068587.1
XM_011511580.2 534 Intron XP_011509882.1
XM_017004628.1 534 UTR 5 XP_016860117.1
XM_017004629.1 534 Intron XP_016860118.1

View Full Product Details