Product Details
- SNP ID
-
hCV972670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:87227342 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTTGGTCACTATGTTCAGTTATT[A/G]GGAGCTTAAAGGAGAAGGACCAGGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUTM2A
PubMed Links
Gene Details
- Gene
- NUTM2A
- Gene Name
- NUT family member 2A
View Full Product Details