Product Details

SNP ID
rs631593
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:201484961 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCAATCTCAATAGTAAAATAGC[A/G]AAGAGGCTCTTGGTTGTACCCTGTA
Phenotype
MIM: 123876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSRP1 PubMed Links

Gene Details

Gene
CSRP1
Gene Name
cysteine and glycine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193570.1 Intron NP_001180499.1
NM_001193571.1 Intron NP_001180500.1
NM_001193572.1 Intron NP_001180501.1
NM_004078.2 Intron NP_004069.1

View Full Product Details