Product Details

SNP ID
rs492660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59176101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTATGTCTTTTAAAGTGGTGA[A/G]AAGTGTCAGATGGAGAGAAAGAGCA
Phenotype
MIM: 616110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTX4 PubMed Links

Gene Details

Gene
DTX4
Gene Name
deltex E3 ubiquitin ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300727.1 Intron NP_001287656.1
NM_015177.1 Intron NP_055992.1
XM_006718483.3 Intron XP_006718546.1

View Full Product Details