Product Details
- SNP ID
-
rs33550
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:112983344 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACAGTTAAAATTTCAGCATTTCAA[C/T]AGAGCGGGCTGAGGAGAACTGGGAA
- Phenotype
-
MIM: 609844
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCP2
PubMed Links
Gene Details
- Gene
- DCP2
- Gene Name
- decapping mRNA 2
View Full Product Details