Product Details

SNP ID
rs186755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123099715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTATGCATGGGCCACTGCACTC[G/T]CTGCGCCGGCTTGTGGGCACCAGCA
Phenotype
MIM: 616982
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRDM6 PubMed Links

Gene Details

Gene
PRDM6
Gene Name
PR domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136239.1 1106 Silent Mutation TCG,TCT S218S NP_001129711.1
XM_011543726.2 1106 Silent Mutation TCG,TCT S18S XP_011542028.1

View Full Product Details