Product Details
- SNP ID
-
rs344586
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:6588220 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAGTGACACCTGGTCCCCTGTCA[C/G]ACACTTAGGCATGATATCATCCACC
- Phenotype
-
MIM: 602840
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD70
PubMed Links
Gene Details
- Gene
- CD70
- Gene Name
- CD70 molecule
View Full Product Details