Product Details

SNP ID
rs27653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60498403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTGAATCCGAGCACAAGTTGA[G/T]TCCCCAGAGTGTAAAACATTGACTC
Phenotype
MIM: 604991
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PART1 PubMed Links

Gene Details

Gene
PART1
Gene Name
prostate androgen-regulated transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details