Product Details

SNP ID
rs185190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60489418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTTTATTTAGTGGTGACATAT[A/T]AACAGTAACAGAAGAGACATTTATG
Phenotype
MIM: 604991 MIM: 600129
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PART1 PubMed Links

Gene Details

Gene
PART1
Gene Name
prostate androgen-regulated transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
PDE4D
Gene Name
phosphodiesterase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104631.1 Intron NP_001098101.1
NM_001165899.1 Intron NP_001159371.1
NM_001197218.1 Intron NP_001184147.1
NM_001197219.1 Intron NP_001184148.1
NM_001197220.1 Intron NP_001184149.1
NM_001197221.1 Intron NP_001184150.1
NM_001197222.1 Intron NP_001184151.1
NM_001197223.1 Intron NP_001184152.1
NM_006203.4 Intron NP_006194.2

View Full Product Details