Product Details

SNP ID
rs216399
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:37298779 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACTAAATAGCAAAGAAACTCTTTA[C/T]TTTACTCGAAAGATCGTTATTTGTC
Phenotype
MIM: 606694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP155 PubMed Links

Gene Details

Gene
NUP155
Gene Name
nucleoporin 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278312.1 Intron NP_001265241.1
NM_004298.3 Intron NP_004289.1
NM_153485.2 Intron NP_705618.1
XM_011514165.2 Intron XP_011512467.1

View Full Product Details