Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243194.1 | 1828 | Intron | NP_001230123.1 | ||
NM_001243195.1 | 1828 | Intron | NP_001230124.1 | ||
NM_014937.3 | 1828 | Intron | NP_055752.1 | ||
XM_006717720.3 | 1828 | UTR 5 | XP_006717783.1 | ||
XM_011539524.2 | 1828 | Intron | XP_011537826.1 | ||
XM_011539525.2 | 1828 | UTR 5 | XP_011537827.1 | ||
XM_011539527.2 | 1828 | Intron | XP_011537829.2 | ||
XM_011539528.2 | 1828 | Intron | XP_011537830.1 | ||
XM_011539529.1 | 1828 | Intron | XP_011537831.1 | ||
XM_017015935.1 | 1828 | Intron | XP_016871424.1 | ||
XM_017015936.1 | 1828 | Intron | XP_016871425.1 |