Product Details

SNP ID
rs2013365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33795911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATGGGGAGGGAATTCACTCCC[A/C]CCTCAACAATTATAGCCAACATTGT
Phenotype
MIM: 616312 MIM: 158270
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LEMD2 PubMed Links

Gene Details

Gene
LEMD2
Gene Name
LEM domain containing 2
There are no transcripts associated with this gene.

Gene
MLN
Gene Name
motilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040109.1 Intron NP_001035198.1
NM_001184698.1 Intron NP_001171627.1
NM_002418.2 Intron NP_002409.1

View Full Product Details