Product Details

SNP ID
rs486652
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:43659970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGGACCTTGGGTCTTGGTCTGT[A/G]AGCCCATTTTCTCCCTGGGTGGCCA
Phenotype
MIM: 609764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014663.2 Intron NP_055478.2
XM_005271354.3 Intron XP_005271411.1
XM_005271355.3 Intron XP_005271412.1
XM_005271356.3 Intron XP_005271413.1
XM_011542467.2 Intron XP_011540769.2
XM_017002912.1 Intron XP_016858401.1

View Full Product Details