Product Details

SNP ID
rs189159450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119088285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGCCCAAAGATGAGAGTGATT[C/T]GCGTGGGTACCCGCAAGAGCCAGGT
Phenotype
MIM: 191350 MIM: 601772 MIM: 609806 MIM: 608549
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPAGT1 PubMed Links

Gene Details

Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
There are no transcripts associated with this gene.

Gene
H2AFX
Gene Name
H2A histone family member X
There are no transcripts associated with this gene.

Gene
HMBS
Gene Name
hydroxymethylbilane synthase
There are no transcripts associated with this gene.

Gene
VPS11
Gene Name
VPS11, CORVET/HOPS core subunit
There are no transcripts associated with this gene.

View Full Product Details