Product Details

SNP ID
hCV14051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48685616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTCCACCCCCACCCCCTTCTTC[T/C]ACACCTTCGCTGGAGATGTAAGTGA
Phenotype
MIM: 300254 MIM: 300392
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SUV39H1 PubMed Links

Gene Details

Gene
SUV39H1
Gene Name
suppressor of variegation 3-9 homolog 1
There are no transcripts associated with this gene.

Gene
WAS
Gene Name
Wiskott-Aldrich syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000377.2 400 Missense Mutation CAC,TAC H115Y NP_000368.1

View Full Product Details