Product Details

SNP ID
rs363387
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:117244053 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCAGACGGCCAGGCCAGTGCACAC[G/T]GCCTCCATCTCAGACAGCTTCCAGA
Phenotype
MIM: 193001
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC18A2 PubMed Links

Gene Details

Gene
SLC18A2
Gene Name
solute carrier family 18 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003054.4 367 Silent Mutation ACG,ACT T68T NP_003045.2

View Full Product Details