Product Details

SNP ID
hCV14902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92213341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTAGGAATTTCCCAGCAATTCT[A/G]TAAGAACATGCGCTGAAGGTGATGC
Phenotype
MIM: 604214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRIT1 PubMed Links

Gene Details

Gene
KRIT1
Gene Name
KRIT1, ankyrin repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194455.1 2898 Nonsense Mutation CAG,TAG Q627* NP_919437.1
NM_194456.1 2898 Nonsense Mutation CAG,TAG Q627* NP_919438.1
Gene
MIR1285-1
Gene Name
microRNA 1285-1
There are no transcripts associated with this gene.

View Full Product Details