Product Details

SNP ID
rs28792
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:135935673 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGGACTTCAAATTTGCTGGAACTTA[A/G]TTGCGTCCCAAGCTGGCTAATACCT
Phenotype
MIM: 609087
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL21 PubMed Links

Gene Details

Gene
FBXL21
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012159.4 430 UTR 5 NP_036291.2

View Full Product Details