Product Details

SNP ID
rs757660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30397148 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGAGTCGGCGATCTGAGCCCCA[A/G]GCAGAAGGAGGCATTGGCCAAGGTG
Phenotype
MIM: 607558
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF215 PubMed Links

Gene Details

Gene
RNF215
Gene Name
ring finger protein 215
There are no transcripts associated with this gene.

Gene
SEC14L2
Gene Name
SEC14 like lipid binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204204.2 208 Missense Mutation AAG,AGG K11R NP_001191133.1
NM_001291932.1 208 UTR 5 NP_001278861.1
NM_012429.4 208 Missense Mutation AAG,AGG K11R NP_036561.1
NM_033382.2 208 Missense Mutation AAG,AGG K11R NP_203740.1

View Full Product Details