Product Details

SNP ID
rs7209877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81670607 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAGCCCTGGGAGTTGGCGAGTC[C/G]GAGCATCCAGCTGGGAGCCAGTGAG
Phenotype
MIM: 614271 MIM: 180073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC137 PubMed Links

Gene Details

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199287.2 641 Intron NP_954981.1
XM_011524738.2 641 Intron XP_011523040.1
XM_017024573.1 641 UTR 3 XP_016880062.1
Gene
OXLD1
Gene Name
oxidoreductase like domain containing 1
There are no transcripts associated with this gene.

Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

View Full Product Details