Product Details

SNP ID
rs11803208
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:204037419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTGCATCACCTTCCTCTCATGCT[A/G]CATCGGATAGTTCCCTGGACAACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details