Product Details
- SNP ID
-
rs35719782
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:58009275 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AAGGAGTTTGCAGATGGGAGCGAAG[C/T]ACCACTGTTGATAATAACCTGAGAG
- Phenotype
-
MIM: 611394
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM110B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7828484] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM110B
- Gene Name
- family with sequence similarity 110 member B
View Full Product Details