Product Details

SNP ID
rs12092624
Assay Type
Validated
NCBI dbSNP Submissions
52
Location
Chr.1:205598862 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGAGCTTCAGTGATGATAGAGAT[C/T]GTCAGAAGGTTCCAGAACACAGAGC
Phenotype
MIM: 600246
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELK4 PubMed Links

Gene Details

Gene
ELK4
Gene Name
ELK4, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001973.3 Intron NP_001964.2
NM_021795.2 Intron NP_068567.1
XM_005244950.4 Intron XP_005245007.1
XM_005244951.4 Intron XP_005245008.1
XM_017000580.1 Intron XP_016856069.1
Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 Intron NP_857595.3
XM_011509229.2 Intron XP_011507531.1

View Full Product Details