Product Details
- SNP ID
-
rs1044385
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:78614399 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCAGTGCACATTAAACAGCATACA[A/T]ACCCATTTTTAAAGACCTATATAGG
- Phenotype
-
MIM: 615906
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RNF219
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114610851] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RNF219
- Gene Name
- ring finger protein 219
- Gene
- RNF219-AS1
- Gene Name
- RNF219 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details