Product Details
- SNP ID
-
rs3922934
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:152626885 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AATTGTATGCAATTTTGGCATAAAT[A/C]TCTGAGAGTCACTGTGGGCCATGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM154
PubMed Links
Gene Details
- Gene
- TMEM154
- Gene Name
- transmembrane protein 154
View Full Product Details