Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020397.3 | Intron | NP_065130.1 | |||
NM_153498.3 | Intron | NP_705718.1 | |||
XM_006717481.3 | Intron | XP_006717544.1 | |||
XM_006717482.3 | Intron | XP_006717545.1 | |||
XM_006717483.3 | Intron | XP_006717546.1 | |||
XM_011519591.2 | Intron | XP_011517893.1 | |||
XM_011519592.2 | Intron | XP_011517894.1 | |||
XM_011519593.2 | Intron | XP_011517895.1 | |||
XM_011519594.2 | Intron | XP_011517896.1 | |||
XM_011519595.2 | Intron | XP_011517897.1 | |||
XM_017016438.1 | Intron | XP_016871927.1 | |||
XM_017016439.1 | Intron | XP_016871928.1 |