Product Details

SNP ID
rs13279448
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104489585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGAGTGTAGCATGATCCCCCCCA[A/G]GAATAAAATATATTGAAAAAATCAC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LRP12 PubMed Links

Gene Details

Gene
LRP12
Gene Name
LDL receptor related protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135703.2 3795 UTR 3 NP_001129175.1
NM_013437.4 3795 UTR 3 NP_038465.1
Gene
MIR548A3
Gene Name
microRNA 548a-3
There are no transcripts associated with this gene.

View Full Product Details