Product Details

SNP ID
rs560494
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168876533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACCAATTGCTTCATGGGAAAATG[G/T]TCTCTTCCTTTAACATAACTATTTC
Phenotype
MIM: 609395
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SPC25 PubMed Links

Gene Details

Gene
SPC25
Gene Name
SPC25, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020675.3 Intron NP_065726.1
XM_011511516.2 Intron XP_011509818.1

View Full Product Details