Product Details

SNP ID
rs713323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48263872 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGCTTTAAAATATGGTTGCTAC[A/G]AACAATGTGACTGAAATAATTTTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4X1 PubMed Links

Gene Details

Gene
OR4X1
Gene Name
olfactory receptor family 4 subfamily X member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004726.1 12 Silent Mutation ACA,ACG T4T NP_001004726.1

View Full Product Details