Product Details

SNP ID
rs734921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45113478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCTGGGAGGTCCTGGGTACTC[A/G]GGGTCAAAGGTCGAGTCAGGTTGCC
Phenotype
MIM: 160993 MIM: 608795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6784 PubMed Links

Gene Details

Gene
MIR6784
Gene Name
microRNA 6784
There are no transcripts associated with this gene.

Gene
NMT1
Gene Name
N-myristoyltransferase 1
There are no transcripts associated with this gene.

Gene
PLCD3
Gene Name
phospholipase C delta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133373.4 2079 Silent Mutation CCC,CCT P652P NP_588614.1
XM_011524253.2 2079 Intron XP_011522555.1

View Full Product Details