Product Details

SNP ID
rs470887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120911805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGAATATAGTTAATTTTAAAAA[A/G]TAGTTATATGGATGTATTATAATTA
Phenotype
MIM: 609381
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STXBP5L PubMed Links

Gene Details

Gene
STXBP5L
Gene Name
syntaxin binding protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308330.1 Intron NP_001295259.1
NM_014980.2 Intron NP_055795.1
XM_006713825.3 Intron XP_006713888.1
XM_011513328.2 Intron XP_011511630.1
XM_011513331.2 Intron XP_011511633.1
XM_011513332.2 Intron XP_011511634.1
XM_011513333.2 Intron XP_011511635.1
XM_017007530.1 Intron XP_016863019.1
XM_017007531.1 Intron XP_016863020.1
XM_017007532.1 Intron XP_016863021.1
XM_017007533.1 Intron XP_016863022.1
XM_017007534.1 Intron XP_016863023.1
XM_017007535.1 Intron XP_016863024.1

View Full Product Details