Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csq-1
Entrez Gene ID
181563
Gene Name
Calsequestrin
Gene Aliases
CELE_F40E10.3
Chromosome Location
Chr.X: 14684877 - 14688327 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078037.7 NP_510438.1 1 - 2 137 90

View Full Product Details