Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lips-8
Entrez Gene ID
185178
Gene Name
LIPaSe related
Gene Aliases
CELE_F31F6.7
Chromosome Location
Chr.X: 14898505 - 14900854 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078073.2 NP_510474.1 2 - 3 376 92

View Full Product Details