Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbg-1
Entrez Gene ID
181595
Gene Name
Rab3 GTPase-activating protein catalytic subunit
Gene Aliases
CELE_F20D1.6
Chromosome Location
Chr.X: 14983533 - 14987376 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078089.5 NP_510490.1 7 - 8 836 58

View Full Product Details