Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flr-4
Entrez Gene ID
181604
Gene Name
Serine/threonine-protein kinase flr-4
Gene Aliases
CELE_F09B12.6
Chromosome Location
Chr.X: 15089397 - 15093585 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078107.6 NP_510508.1 6 - 7 851 94
GenBank mRNA AB012700.1 - 6 - 7 873 94

View Full Product Details