Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jud-4
Entrez Gene ID
182109
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C02D4.1
Chromosome Location
Chr.X: 15284115 - 15286484 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078133.6 NP_510534.3 4 - 5 509 65

View Full Product Details