Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glb-28
Entrez Gene ID
189438
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_Y15E3A.2
Chromosome Location
Chr.X: 15314874 - 15317246 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078136.3 NP_510537.1 1 - 2 96 70

View Full Product Details