Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
csb-1
Entrez Gene ID
181674
Gene Name
human CSB (Cockayne Syndrome B) homolog
Gene Aliases
CELE_F53H4.1
Chromosome Location
Chr.X: 15861294 - 15865918 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078206.5 NP_510607.2 7 - 8 2202 61

View Full Product Details