Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sgk-1
Entrez Gene ID
181697
Gene Name
protein-coding
Gene Aliases
CELE_W10G6.2
Chromosome Location
Chr.X: 16254843 - 16262185 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047834.4 NP_001041299.1 8 - 9 774 88
NM_001381177.1 NP_001367134.1 8 - 9 908 88

View Full Product Details