Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
let-2
Entrez Gene ID
181708
Gene Name
protein-coding
Gene Aliases
CELE_F01G12.5
Chromosome Location
Chr.X: 16377600 - 16389371 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078262.6 NP_510663.1 11 - 12 1372 71
NM_078263.4 NP_510664.1 11 - 12 1387 71

View Full Product Details