Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-11
Entrez Gene ID
181745
Gene Name
Receptor-type guanylate cyclase gcy-11
Gene Aliases
CELE_C30G4.3
Chromosome Location
Chr.X: 17046241 - 17055581 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373625.3 NP_001359843.1 21 - 22 3149 74

View Full Product Details